Stampa…

Acido 4-indolil-3-butirrico, powder_off-white

Fornitore: MP Biomedicals
Pericolo

Synonyms: 3-Indolebutyric acid, Indole-3-butyric acid

0210204201 0210204205 0210204225 0210204250
ICNA0210204201EA 33.4 EUR
ICNA0210204201 ICNA0210204205 ICNA0210204225 ICNA0210204250
Acido 4-indolil-3-butirrico, powder_off-white
Acido 4-indolil-3-butirrico

Indole-3-butyric acid is used as plant growth hormone.


A naturally occurring phytohormone auxin (plant growth regulator). Promotes root formation in cuttings, but does not affect ethylene levels.


Attenzione: Research Use Only (RUO).

Formula: C₁₂H₁₃NO₂
Peso molecolare: 203.24 g/mol
Densità: 1.27
MDL Number: MFCD00005664
CAS Number: 133-32-4
UN: 2811
ADR: 6.1 III
Merck Index: 13,04987

Visualizza i codici

Risultati del test di specificazione

Identity Test Passes
FTIR Conforms to Standard
Melting Point 118 - 126 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR