Stampa…

D(-)Arabinosio ≥99%, in polvere

Fornitore: MP Biomedicals

Synonyms: D-(-)-Arabinose, (2S,3R,4R)-2,3,4,5-Tetrahydroxypentanal

0210069991 0210069925 0210069980
ICNA0210069991EA 1912 EUR
ICNA0210069991 ICNA0210069925 ICNA0210069980
D(-)Arabinosio ≥99%, in polvere
D(-)Arabinosio

D-( -)-Arabinose is a D-ribose analogue present in sugar polymers such as the arabinogalatans of mycobacterial cell walls and plant tissues. It is a substrate used to identify, differentiate and characterise arabinose isomerase(s) that have commercial value in the production of tagatose, a low calorie sweetener.


D-Arabinose is a reducing sugar. It is a pentose analog of D-ribose that is a constituent of mycobacterial cell wall arabinogalactans. It is also a substrate for D-erythroascorbic acid synthesis in yeast.


Soluble in water (50 mg/ml) giving clear, colourless to faint yellow solution.

Formula: C₅H₁₀O₅
Punto di fusione: 160 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00135608
CAS Number: 10323-20-3
EINECS: 233-708-5

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR