Stampa…

Nicotinammide adenina dinucleotide (forma ossidata) ≥98%

Fornitore: MP Biomedicals

02100499.1 0210049901 0210049905 0210049910
ICNA02100499.1EA 53.2 EUR
ICNA02100499.1 ICNA0210049901 ICNA0210049905 ICNA0210049910
Nicotinammide adenina dinucleotide (forma ossidata) ≥98%
Nicotinammide adenina dinucleotide (forma ossidata)

Major electron acceptor molecule in biological oxidations. Acts as a coenzyme for hydride-transfer enzymes and a substrate for NAD(+)-consuming enzymes, which include ADP-ribose transferases, poly(ADP-ribose) polymerases, cADP-ribose synthases and sirtuins. Recently reported to be a P2Y11 agonist.


  • Lyophilised white powder
  • High purity


Storage temperature: –20 °C


pH: 2 to 4 (1% aquous solution)

Formula: C₂₁H₂₇N₇O₁₄P₂
MDL Number: MFCD00036253
CAS Number: 53-84-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR