Stampa…

3-Indolyl-ß-D-galactopyranoside, Sigma-Aldrich®

Fornitore: SIGMA ALDRICH MICROSCOPY
Attenzione


M-Clarity™ quality level = MQ200

I7513-5MG
I7513-5MGEA 89.6 EUR
I7513-5MG
3-Indolyl-ß-D-galactopyranoside, Sigma-Aldrich®
3-Indolyl-ß-D-galactopyranoside
Formula: C₁₄H₁₇NO₆
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00083389
CAS Number: 126787-65-3
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36a40e15ec6e6c0c555c76abde2096e2c693e088&vid=36a40e15ec6e6c0c555c76abde2096e2c693e088&prid=web&clid=SAPDAM

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR