Stampa…

Indocianina verde, Sigma-Aldrich®

Fornitore: SIGMA ALDRICH MICROSCOPY
Attenzione


M-Clarity™ quality level = MQ200

I2633-100MG I2633-25MG I2633-50MG
I2633-100MGEA 273 EUR
I2633-100MG I2633-25MG I2633-50MG
Indocianina verde, Sigma-Aldrich®
Indocyanine green

A negatively charged polymethine dye that forms noncovalent fluorescent complexes with proteins.

Formula: C₄₃H₄₇N₂NaO₆S₂
Temperatura di stoccaggio: Room Temperature
MDL Number: MFCD00013078
CAS Number: 3599-32-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59d432a05d02412effb6464eaca953d16e959a1b&vid=6002e8e16ac6c957b5906d060900607e4641ec48&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance (Colour) Green to very dark green and brown to very dark brown
Appearance (Form) Powder
Solubility (Colour) 0.01 g in 10 ml water
Solubility (Method) 0.01 g in 10 ml water
Nitrogen Content 2.7 - 3.6 %
Dye Content ≥75 %
Recommended Retest Period 72 Months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR