Stampa…
Snap caps, 11 mm, level 3 high performance applications, SureSTART™
Fornitore: Thermo Fisher Scientific
6PRC11ST1 6PRC11ST101R 6PRC11ST101RF 6PRC11ST101RT 6PRC11ST1F 6PRC11ST1R 6PRC11ST1RF 6PRC11ST1RT 6PRC11ST1T 6PRC11STS1 6PRC11STS1F 6PRC11STS1R 6PRC11STS1RF 6PRC11STS1RT 6PRC11STS1T 6PRC11STS1X 6PRC11STS1XF 6PRC11STS1XT 6PRC11ST101 6PRC11ST101F 6PRC11ST101T
HYPE6PRC11ST1EA 39.5 EUR
HYPE6PRC11ST1 HYPE6PRC11ST101R HYPE6PRC11ST101RF HYPE6PRC11ST101RT HYPE6PRC11ST1F HYPE6PRC11ST1R HYPE6PRC11ST1RF HYPE6PRC11ST1RT HYPE6PRC11ST1T HYPE6PRC11STS1 HYPE6PRC11STS1F HYPE6PRC11STS1R HYPE6PRC11STS1RF HYPE6PRC11STS1RT HYPE6PRC11STS1T HYPE6PRC11STS1X HYPE6PRC11STS1XF HYPE6PRC11STS1XT HYPE6PRC11ST101 HYPE6PRC11ST101F HYPE6PRC11ST101T
Snap caps, 11 mm, level 3 high performance applications, SureSTART™
Ghiere Fiale per cromatografia
Use Thermo Scientific™ SureSTART™ 11 mm Snap caps with snap vials that have an 11 mm opening, including our SureSTART 2 ml glass snap Vials, 1.5 ml total recovery glass snap vials, high recovery glass snap vials, and glass snap micro vials for <2 ml samples (level 3).
Select level 3 products for ultimate performance and sensitivity from your methods, while maintaining the highest levels of sample security and integrity. Choose from soft polyethylene caps with double snap functionality that guarantees a tight seal.
These snap caps include all-purpose silicone/PTFE septa of variousMore Product Information