Stampa…
Serum-free media, HyClone™ SFMTranfx-293 media
Fornitore: HyClone products (Cytiva)
SH30860.02 SH30860.03 SH30860.04 SH30861.01
HYCLSH30860.02EA 86.9 EUR
HYCLSH30860.02 HYCLSH30860.03 HYCLSH30860.04 HYCLSH30861.01
Serum-free media, HyClone™ SFMTranfx-293 media
Terreni per colture cellulari
HyClone™ SFMTransfx-293 is a serum-free, animal derived component-free medium designed to support the growth of HEK 293 cultures and promote transfection using lipofection or similar methods. Manufactured according to cGMP guidelines.
  • Supports high transfection efficiency
  • Metabolically designed for high cell yield and recombinant protein/vector production
  • Allows for direct or sequential adaptation
HyClone SFMTransfx-293 Media is a regulatory-friendly medium developed through the Metabolic Pathway Design to support high transfection efficiency, productivity and cell density in suspension cultures.