Stampa…
Quick-Disconnect Coupler, M16x1
Fornitore: HUBER
10790
HUBE10790EA 0 EUR
HUBE10790
Quick-Disconnect Coupler, M16x1
Raccordi e connettori
Note: When using ethanol as heat transfer fluid minimum working temperature limit −60 °C.