Stampa…
Multi-position magnetic stirrers, Variomag Biosystem
Fornitore: THERMO H + P LABORTECHNIK
50118919 50118915 50118918 50119113 50119114 50118917
HPLT50118919EA 1530 EUR
HPLT50118919 HPLT50118915 HPLT50118918 HPLT50119113 HPLT50119114 HPLT50118917
Multi-position magnetic stirrers, Variomag Biosystem
Dispositivi agitatori Agitatori magnetici, multiposizione
Single and multi-position stirrers for up to four vessels. Biosystem models and accessory Biomodul 40 B controller are suitable for mixing culture broths, micro-carrier cultures and cell suspensions. Units provide slow and even stirring to protect cultures.
  • Rugged construction with hermetically sealed stainless steel housing, units resist agressive media and can be cleaned under running water
  • Biosystem stirrers generate little to no heat so cultures can be gently stirred, avoiding potential thermal damage
  • Automatic start up for safe stirrer acceleration