Stampa…
Chromatography data station, DataApex ClarityVA
Fornitore: COLLECTION PREFIX HPLC
DAAPC40 DAAPC50 DAAPC59 DAAPC5918 DAAPC620 DAAPC50-18B1 DAAPC50-18B2 DAAPC50-18B3 DAAPU31 DAAPU32 DAAPU34 DAAPC50-18 DAAPC50-18B4 DAAPC50-18B4AC DAAPC50-18B5 DAAPC50-18B5AC
906-0162EA 1890 EUR
906-0162 906-0161 906-0163 906-0189 906-0190 906-0192 906-0193 906-0197 906-0194 906-0195 906-0196 HPLCDAAPC50-18 HPLCDAAPC50-18B4 HPLCDAAPC50-18B4AC HPLCDAAPC50-18B5 HPLCDAAPC50-18B5AC
Chromatography data station, DataApex ClarityVA
Software Chromatography Software
ClarityVA is a flexible and powerful Chromatography Data System (CDS) that controls HPLC and GC systems. This modular software includes a lot of controls for many different systems, modules and interfaces. The flexibility enables an easy control of HPLC systems in combination with a user friendly surface.
  • Direct control of Hitachi HPLC (Chromaster and Primaide)
  • Modules for specific calculation and methodology
  • Tools for regulated environment
  • Extended user support
  • Language localisations (English, German, French,...)
ClarityVA is useable for modernMore Product Information