Stampa…
Medical Masks, Type II Tie-On
Fornitore: Medicom
M24101-30
HOPEM24101-30EA 72.7 EUR
HOPEM24101-30
Medical Masks, Type II Tie-On
Maschere Mascherine Chirurgiche
Medical Device: Class I - Medical mask type: Type II
  • Protects the environment from droplets emitted by the wearer
  • Effective against bacteria, particles, and viruses (BFE, PFE, & VFE)
  • Kolmi® masks feature the exclusive Origami folding for a better comfort and a perfect fit to the wearer's face
  • For maximum comfort, the inner layer is designed with a material limiting facial irritation.
  • With Feeldry technology, the inner layer has been specially designed to absorb sweat and thus avoid sensation moisture inside the mask
  • Hypoallergenic
Medical masks