Stampa…

Clopidol Analytical standard, reference material, Fluka™

Fornitore: Honeywell Chemicals

R1803-100MG
HONCR1803-100MGEA 123.31 EUR
HONCR1803-100MG
Clopidol Analytical standard, reference material, Fluka™
Standard Standard organici

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.


  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery

Visualizza i codici

Specifiche

Descrizione Clopidol
Environmentally Preferable
Forma fisica Solid
CAS No. 2971-90-6
N. MDL MFCD00867211
Peso molecolare 192.04 g/mol
Punto di fusione 320 °C
Condizioni di conservazione RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR