Stampa…

Pesticide standard, Tebufenozide, reference material, Honeywell Fluka™

Fornitore: Honeywell Chemicals

R1619-100MG
HONCR1619-100MGEA 203.69 EUR
HONCR1619-100MG
Pesticide standard, Tebufenozide, reference material, Honeywell Fluka™
Standard Standard organici

High quality organic reference materials produced according to ISO 9001.


Informazioni per l'ordine: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.

Visualizza i codici

Specifiche

Descrizione Tebufenozide
Environmentally Preferable
CAS No. 112410-23-8
Formula C₂₂H₂₈N₂O₂
Temperatura di conservazione Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR