Stampa…

Analytical standard, Monuron-dimethyl, reference material, Honeywell Fluka™

Fornitore: Honeywell Chemicals

R1047-10MG
HONCR1047-10MGEA 532.85 EUR
HONCR1047-10MG
Analytical standard, Monuron-dimethyl, reference material, Honeywell Fluka™
Standard Standard organici

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Visualizza i codici

Specifiche

Descrizione Monuron-dimethyl
Environmentally Preferable
CAS No. 217488-65-8
Formula C₉D₆H₅ClN₂O
Temperatura di conservazione Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR