Stampa…

Analytical standard, Reserpine-d₉, reference material, Honeywell Fluka™

Fornitore: Honeywell Chemicals

R1000-10MG
HONCR1000-10MGEA 693.62 EUR
HONCR1000-10MG
Analytical standard, Reserpine-d₉, reference material, Honeywell Fluka™
Standard Standard organici

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Visualizza i codici

Specifiche

Descrizione Reserpine-d₉
Environmentally Preferable
CAS No. 84759-11-5
Formula C₃₃D₉H₃₁N₂O₉
Punto di fusione 273…276 °C
Temperatura di conservazione Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR