Stampa…
Potassium Standard for Ion Chromatography, (CRM), Fluka™
Fornitore: Honeywell Chemicals
E3041-100ML
HONCE3041-100MLEA 55.15 EUR
HONCE3041-100ML
Potassium Standard for Ion Chromatography, (CRM), Fluka™
Standard Standard cationici Standard di potassio
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Certificazioni: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).