Stampa…
Concentrated Eluent for Ion Chromatography, Sodium Carbonate/Sodium Bicarbonate Solution 2
Fornitore: Honeywell Chemicals
E2208-500ML
HONCE2208-500MLEA 233.41 EUR
HONCE2208-500ML
Concentrated Eluent for Ion Chromatography, Sodium Carbonate/Sodium Bicarbonate Solution 2
Standard Standard anionici Standard multi-elemento anionico
Certified Reference Material (CRM).
0.18 M Na₂CO₃ / 0.17 M NaHCO₃ in water