Stampa…

Rosso Clorofenolo soluzione for PANPEHA indicator paper, Riedel-de Haën™

Fornitore: Honeywell Chemicals

34658-5L
HONC34658-5LEA 1561.81 EUR
HONC34658-5L
Rosso Clorofenolo soluzione for PANPEHA indicator paper, Riedel-de Haën™
Rosso Clorofenolo
Formula: C₁₉H₁₂Cl₂O₅S
MDL Number: MFCD00005877
CAS Number: 4430-20-0

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR