Stampa…

Urea ≥99.5%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™

Fornitore: Honeywell Chemicals

33247H-1KG 33247H-2.5KG 33247H-5KG
HONC33247H-1KGEA 89.87 EUR
HONC33247H-1KG HONC33247H-2.5KG HONC33247H-5KG
Urea ≥99.5%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Urea
Formula: NH₂CONH₂
Punto di fusione: 133…134 °C
Densità: 1,32 g/cm3 (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Identity complying
Assay Min. 99.5 %
Assay (calculated to the dried substance) 99 - 101 %
Melting range 132 - 135 °C
Water insoluble matter Max. 0.01 %
Loss on drying (105°C, 1 h) Max. 1.0 %
Sulfated ash Max. 0.01 %
Ammonium (NH₄) Max. 0.05 %
Alkalinely reacting substances complying
Iron (Fe) Max. 0.0002 %
Heavy metals (as Pb) Max. 0.0005 %
Chloride (Cl) Max. 0.0005 %
Sulfate (SO₄) Max. 0.001 %
Biuret (carbamylurea) Max. 0.1 %
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR