Stampa…
MacConkey agar with CV, NaCl, and 0,15% Bile Salts, dehydrated culture media
Fornitore: HIMEDIA
M081A-500G
HIMEM081A-500GEA 97.6 EUR
HIMEM081A-500G
MacConkey agar with CV, NaCl, and 0,15% Bile Salts, dehydrated culture media
Terreni per microbiologia Terreni di coltura per microbiologia, disidratati
Used to identify Enterobacteriaceae in the presence of coliforms and lactose nonfermenters from water, sewage, and food products.
  • Microbiological culture media
  • 500 g poly bottles