Stampa…
V.C.N. Supplement
Fornitore: HIMEDIA
FD023-5X5VL
HIMEFD023-5X5VLEA 0 EUR
HIMEFD023-5X5VL
V.C.N. Supplement
Terreni per microbiologia Additivi e supplementi per microbiologia
An antibiotic supplement, recommended for the selective isolation of N. gonorrhoeae and N. meningitidis.
  • Store at 2 to 8 °C