Stampa…
Avantor® ACE® HILIC-B, HPLC Columns, 5 µm
Fornitore: Avantor
HILB-5-3030EA 0 EUR
HILB-5-3030 HILB-5-3020 HILB-5-2530 HILB-5-7530 HILB-5-7520 HILB-5-1530 HILB-5-1520 HILB-5-1510 HILB-5-1220 HILB-5-1030 HILB-5-1020 HILB-5-1010 HILB-5-0530 HILB-5-0520 HILB-5-0510 HILB-5-2520
Avantor® ACE® HILIC-B, HPLC Columns, 5 µm
Colonne cromatografiche
Avantor® ACE® HILIC-B columns have been designed specifically for HILIC applications and are especially useful for the separation of polar analytes. These columns are part of a suit of 3 proprietary HILIC columns (HILIC-A, HILIC-B and HILIC-N) developed to enable chromatographers to achieve reproducible and successful HILIC separations using a systematic approach to method development.
  • Proprietary aminopropyl basic character HILIC phase
  • Compatible with all manufacturers HPLC/UHPLC systems
  • Excellent column performance, inertness and robustness
  • Simplified