Stampa…
Avantor® Alltima HP, HPLC Columns, C18-AQ, 3 µm
Fornitore: Avantor
HICH87816EA 538 EUR
HICH87816 HICH87832 HICH87807 HICH87808
Avantor® Alltima HP, HPLC Columns, C18-AQ, 3 µm
Colonne cromatografiche
Avantor® Alltima HP columns combine a range of different phase chemistries with high purity silica. Avantor® Alltima HP columns contain the highest carbon load for superior retention and loadability.
  • High purity silica
  • Excellent column stability
  • Low to no detectable column bleed
  • Multiple selectivity options
These columns are 100% water wettable for applications requiring high aqueous mobile phases.
These columns are combining the selectivity and performance needed to overcome the most challenging separation needs. The low column bleed makes theseMore Product Information