Stampa…
Avantor® Hichrom HI-5, GC Columns
Fornitore: Avantor
HI02-05-005-2EA 444 EUR
HI02-05-005-2 HI02-05-005-2.5 HI02-10-010-5 HI02-10-020-15 HI02-10-040-10 HI02-10-040-15 HI02-15-015-10 HI02-15-015-30 HI02-25-025-15G5 HI02-25-025-30G5 HI02-25-025-60G5 HI02-25-100-30G5 HI02-32-100-30G5 HI02-53-050-30G10 HI02-53-100-30G5 HI02-18-018-10 HI02-18-018-20 HI02-18-018-40 HI02-18-018-50 HI02-18-018-60 HI02-18-020-40 HI02-20-010-20 HI02-20-010-50 HI02-20-020-50 HI02-20-020-60 HI02-20-100-20 HI02-20-100-40 HI02-20-100-50 HI02-25-010-50 HI02-25-025-105 HI02-25-025-30 HI02-25-025-60 HI02-25-050-10 HI02-25-050-105 HI02-25-050-15 HI02-25-050-30 HI02-25-050-60 HI02-25-100-105 HI02-25-100-30 HI02-32-010-10 HI02-32-010-105 HI02-32-010-15 HI02-32-010-30 HI02-32-025-10 HI02-32-025-105 HI02-32-025-15 HI02-32-025-30 HI02-32-025-60 HI02-32-050-105 HI02-32-050-50 HI02-32-050-60 HI02-32-100-10 HI02-32-100-105 HI02-32-100-30 HI02-32-100-50 HI02-32-150-10 HI02-32-150-105 HI02-32-150-50 HI02-32-150-60 HI02-32-300-105 HI02-32-300-15 HI02-32-300-50 HI02-32-500-105 HI02-53-025-15 HI02-53-025-60 HI02-53-050-50 HI02-53-100-15 HI02-53-100-50 HI02-53-100-60 HI02-53-100-75 HI02-53-150-15 HI02-53-150-75 HI02-53-300-15 HI02-53-300-50 HI02-53-300-60 HI02-53-300-75 HI02-53-500-60 HI02-53-500-75 HI02-53-700-15 HI02-53-700-30 HI02-32-500-25 HIGCHI0232025155IN HIGCHI02-32-050-25 HIGCHI02-32-017-25 HIGCHI02-32-012-50 HIGCHI02-32-052-50
Avantor® Hichrom HI-5, GC Columns
Colonne cromatografiche
Really versatile general purpose low-polarity phase for a wide range of applications, including (for example): PCBs, essential oils, pesticides, semivolatiles and many more. HI-5 is a crossbonded phase and assure a long lifetime even under high temperature limits.
  • Phase composition: Crossbonded 5% phenyl - 95%Methyl Polysiloxane
  • Long lifetime
Max temperature limits may change depending on the film thickness.