Stampa…

Ematossilina (Harris), Sigma-Aldrich®

Fornitore: SIGMA ALDRICH MICROSCOPY

Synonyms: DcH-48, Natural Black I, Hydroxybrasilin

HHS16-500ML
HHS16-500MLEA 57.6 EUR
HHS16-500ML
Ematossilina (Harris), Sigma-Aldrich®
Ematossilina (soluzione colorante)

General purpose nuclear stain, regressive type. Used with hematoxylin and eosin staining.

Formula: C₁₆H₁₄O₆
Peso molecolare: 302.28 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00078111
CAS Number: 517-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ae0e0602b7d723ea9208c209f429d857cb65261&vid=ebfed848ebd37a09c3b3eb1647f5570c4320e175&prid=web&clid=SAPDAM IVDLogo

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR