Stampa…
Capillary columns, DB-WAXetr
Fornitore: Agilent
122-7332E 122-7333E 122-7362E 123-7332E 123-7354E 123-7363E 125-7332E 125-7334E
HEWL122-7332EEA 0 EUR
HEWL122-7332E HEWL122-7333E HEWL122-7362E HEWL123-7332E HEWL123-7354E HEWL123-7363E HEWL125-7332E HEWL125-7334E
Capillary columns, DB-WAXetr
Colonne cromatografiche
DB-WAXetr is a high polarity polyethylene glycol (PEG) column for the analysis of alcohols, solvents, benzenes, styrenes, and organic acids, among others.
  • Excellent column-to-column repeatability
  • Fast and accurate operation within temperature limits of 30 to 260/280 °C
  • Equivalent to USP Phase G16
  • High polarity
  • Bonded, cross-linked and solvent rinsable
Similar phases: SUPELCOWAX 10, SUPEROX II, CB-WAX, Stabilwax, BP-20, 007-CW, Carbowax, Rtx-WAX, ZB-WAX, ZB-WAX plus.