Stampa…
Laboratory Freezer, ES
Fornitore: Thermo Scientific
98F-AEV-TS
HERA98F-AEV-TSEA 1210 EUR
HERA98F-AEV-TS
Laboratory Freezer, ES
Congelatori Congelatori per usi generici/di laboratorio
Thermo Scientific™ ES Series Lab Freezers, designed for general-purpose lab storage and routine sample protection.
  • Integrated controller
  • Digital display
  • High- and low-temperature alarms
  • Standard door locks
  • Low energy consumption
  • Access ports
  • CFC-free, HCFC-free
Cabinet material: Scratch-resistant painted steel