Stampa…
Herasafe™ 2030i CTS™ Biological Safety Cabinets
Fornitore: Thermo Scientific
51033758 51033759 51033760 51033761
HERA51033758EA 21800 EUR
HERA51033758 HERA51033759 HERA51033760 HERA51033761
Herasafe™ 2030i CTS™ Biological Safety Cabinets
Cappe e involucri Armadi di sicurezza per biologia
Thermo Scientific™ Herasafe™ 2030i Biological Safety Cabinet Cell Therapy Systems (CTS™) Series consists of equipment, documentation and certifications to support your Good Manufacturing Practice (GMP) needs.
  • Ease-of-use and ergonomics
  • Smart protection
  • Standard USB data output
  • Intuitive touchscreen interface for clear indication of status and performance
  • Ergonomical armrest and optional autoclavable padded versions
  • Interior LED lights and side windows
  • Self-adjusting germicidal UV light intensity
Compliance services are available to help ensure aMore Product Information