Stampa…
Cuvette rack
Fornitore: HELLMA
3001.10
HELL3001.10EA 33.1 EUR
HELL3001.10
Cuvette rack
Rack Rack portacuvette
Polypropylene.
Rack for 16 square cells with 10 mm path length with inclined profile to aid cell handling.