Stampa…
Chillers, Hei-CHILL 700 Pro
Fornitore: Heidolph Instruments GmbH & Co.KG
591-01650-00
HEID591-01650-00EA 4970 EUR
HEID591-01650-00
Chillers, Hei-CHILL 700 Pro
Termostati a circolazione Sistemi a circolazione chiusa Termostati a circolazione raffreddanti
Powerful 700 W cooling for every evaporation process and large volumes.
  • High performance for achieving the target temperature quickly and for keeping values constant even when using high-polar solvents
  • Environmentally friendly by using natural refrigerants (R-290) and an efficient cooling system with low-energy fans
  • Touchscreen controller with glass surface and integrated level indicator
Reduced running noises for a comfortable work thanks to speed-controlled fans in line with your requirements. For applications from −10 to +40 °C with a temperatureMore Product Information