Stampa…
Rotary Evaporators, Hei-VAP Expert Control
Fornitore: Heidolph Instruments GmbH & Co.KG
574-02610-00 574-02315-00 573-02310-00 573-02100-00 573-02300-00 573-02305-00 573-02500-00 573-02600-00 573-02110-00 573-02315-00 573-02510-00 573-02610-00 574-02100-00 574-02300-00 574-02305-00 574-02500-00 574-02600-00 574-02110-00 574-02310-00 574-02510-00 574-22600-00 574-23600-00 574-29300-00 574-37300-00 574-38310-00 574-32300-00 574-33300-00
HEID574-02610-00EA 8530 EUR
HEID574-02610-00 HEID574-02315-00 HEID573-02310-00 HEID573-02100-00 HEID573-02300-00 HEID573-02305-00 HEID573-02500-00 HEID573-02600-00 HEID573-02110-00 HEID573-02315-00 HEID573-02510-00 HEID573-02610-00 HEID574-02100-00 HEID574-02300-00 531-1468 HEID574-02500-00 HEID574-02600-00 HEID574-02110-00 HEID574-02310-00 HEID574-02510-00 HEID574-22600-00 HEID574-23600-00 HEID574-29300-00 HEID574-37300-00 HEID574-38310-00 HEID574-32300-00 HEID574-33300-00
Rotary Evaporators, Hei-VAP Expert Control
Evaporatori Evaporatori rotanti
Hei-VAP Expert Control rotary evaporators allow central control of all process parameters: Vacuum, cooling temperature, rotation and heating bath temperature. The units have separate knobs for quick access: Right for direct vacuum control, the left one for rotation has a lock function to prevent accidental adjustment
  • 5" touch display for controlling the entire system including the vacuum and the cooling temperature
  • For precise monitoring, the pictogram on the display also illustrates active functions
  • Set values either via the touch display or the separate