Stampa…
Magnetic Stirrers, Hei-PLATE Mix 'n' Heat Expert
Fornitore: Heidolph Instruments GmbH & Co.KG
506-31100-00 506-32100-00
HEID506-31100-00EA 1590 EUR
HEID506-31100-00 HEID506-32100-00
Magnetic Stirrers, Hei-PLATE Mix 'n' Heat Expert
Dispositivi agitatori Agitatori magnetici
The magnetic stirrer Hei-PLATE Mix 'n' Heat Expert convinces with its intuitive handling. You have an easily legible eye on all values and settings at any time.
The handling is made via two push-and-rotary controls with LED-rings for an easy identification of the active functions. Separate timers for heating and stirring functions create flexibility and let you control your experiments better. The user-friendly menu navigation makes working with this device particularly easy.
For controlling the temperature, you can choose between three heating modes: FAST for a particularly quickMore Product Information