Stampa…
Microscopy slide according to Undritz
Fornitore: GLASWARENFABRIK KARL HECHT
40619010
HECH40619010EA 5 EUR
HECH40619010
Microscopy slide according to Undritz
Vetrini Vetrini di controllo per microscopia
This microscopy slide is designed for blood smears according to Undritz.
  • Polished surface
  • Clear glass