Stampa…
Haematocrit sealing material
Fornitore: GLASWARENFABRIK KARL HECHT
40566010
HECH40566010EA 3.6 EUR
HECH40566010
Haematocrit sealing material
Guarnizioni Composti sigillanti
This sealing material prevents the draining of blood samples. For sealing several hundred micro hematocrit capillaries.