Stampa…
Water distilling apparatus
Fornitore: GLASWARENFABRIK KARL HECHT
40225010
HECH40225010EA 1810 EUR
HECH40225010
Water distilling apparatus
Purificatori d'acqua Distillatori
Complete water distilling apparatus with heater and directions for use.
  • With special outlet for practical emptying and cleaning
  • Without tubing
Required quantity of cooling water: approx. 45 l/h
Temperature of cooling water (discharge): max. +60 °C/+140 °F
Water quality
Conductivity: 0,85 µs
pH: 7,06