Stampa…
Reagents for single parameter colorimeter HI-93701 chlorine (total)
Fornitore: Hanna
HI93701-T
HANNHI93701-TEA 88.6 EUR
HANNHI93701-T
Reagents for single parameter colorimeter HI-93701 chlorine (total)
Kit per analisi ambientali
Total chlorine liquid reagents are used for the colorimetric determination of total and free chlorine, for use with Hanna’s compatible benchtop and portable photometers.
  • Pre-made reagents for ease of use
  • Prepared with high purity chemicals
  • Marked with expiration date and lot number for traceability
  • 300 tests free or total chlorine
Chlorine liquid reagents (A-C) are high quality liquid reagents that allow for users to achieve fast and accurate colorimetric measurements. The methods use a DPD reagent in which the reaction between chlorine and reagent causes a pink tint in the sample.