Stampa…
Baths and circulators, stainless steel, SAHARA PPO S21P series
Fornitore: THERMO ELECTRON LED
1563211 1553211 1523211 1533211
HAKA1563211EA 3050 EUR
HAKA1563211 HAKA1553211 HAKA1523211 HAKA1533211
Baths and circulators, stainless steel, SAHARA PPO S21P series
Termostati a circolazione Sistemi a circolazione aperta Bagni con circolatori rimovibili
These heated bath circulators maintains the ambient temperature. The SAHARA series of heated bath circulators are available with a wide variety of work area dimensions to meet internal or external circulation requirements.
  • A selection of different controllers will create the perfect fit for application
  • Rugged and corrosion-resistant for high temperature applications up to 200 °C
  • Modified polyphenylene oxide baths are thermally resistant up to 100 °C
  • Deliver exceptional temperature performance with operational savings
  • Recommended for calibration,