Stampa…
Portable incubator
Fornitore: Hach
2569900
HACH2569900EA 2560 EUR
HACH2569900
Portable incubator
Incubatori Incubatori da banco compatti
Small and lightweight, the unit is ideal for reliable, accurate incubation for total coliform, fecal coliform and E. coli testing. The incubator can be used for Presence/Absence (P/A), Membrane Filtration (MF) and Most Probable Number (MPN) procedures.
  • Protable, designed for field use in Microbiological Environmental Laboratories (MELs)
  • Can be used with a variety of power sources (12 Vdc via vehicle cigarette lighter socket connection, 12 V DC rechargeable battery or 115/230 V AC Battery Eliminator)
Informazioni per l'ordine: Supplied withoutMore Product Information