Stampa…
Lauryl tryptose broth tubes
Fornitore: Hach
2162315
HACH2162315EA 63 EUR
HACH2162315
Lauryl tryptose broth tubes
Terreni per microbiologia Terreni pronti per microbiologia
Lauryl tryptose broth tubes, single strength, selectivity: total coliform bacteria (presumptive).
  • Save time and labour costs with ready to use MPN tubes, with minimal training requirements