Stampa…
Mixing measuring cylinders, tall form, class B, glass
Fornitore: Hach
2088640
HACH2088640EA 61 EUR
HACH2088640
Mixing measuring cylinders, tall form, class B, glass
Cilindri Cilindri di misurazione
Glass cylinder, Class B.
  • Calibrated 'to contain' (TC)
  • Molded hexagonal base
  • Glass stopper
  • Graduations in durable white enamel