Stampa…

Ferroina (1,10-Fenantrolina-ferro solfato-complesso) soluzione Indicatore, for COD determination (low range)

Fornitore: Hach

2055133
HACH2055133EA 38 EUR
HACH2055133
Ferroina (1,10-Fenantrolina-ferro solfato-complesso) soluzione Indicatore, for COD determination (low range)
Ferroina (1,10-Fenantrolina-ferro solfato-complesso)

Oxidation-reduction indicator solution used in determination of COD, low range, by ferrous titration.

Formula: C₃₆H₂₄FeN₆SO₄
MDL Number: MFCD00036428
CAS Number: 14634-91-4

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR