Stampa…
Potassium iodide-iodate standard solution, 0,0125 N
Fornitore: Hach
1400153
HACH1400153EA 95.5 EUR
HACH1400153
Potassium iodide-iodate standard solution, 0,0125 N
Standard Standard per analisi ambientali
Used for determination of sulphite by burette iodometric titration.