Stampa…

O-Methyl-L-tyrosine 98%

Fornitore: Thermo Fisher Scientific

H63096.03 H63096.06
H63096.03EA 43.1 EUR
H63096.03 H63096.06
O-Methyl-L-tyrosine 98%
O-Methyl-L-tyrosine

Purity: 98% CAS No.: 6230-11-1 Molecular Formula: C10H13NO3 Formula Weight: 195.22 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 259-261° dec Beilstein Reference: 2212726 Optical Rotation: -7°(c=1 in 1M HCl) MDL No.: MFCD00002604

Formula: C₁₀H₁₃NO₃
Punto di fusione: 259…261 °C (dec.)
MDL Number: MFCD00002604
CAS Number: 6230-11-1
EINECS: 228-333-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR