Stampa…

3,4-Dihydro-2H-pyran-2-methanol 96%

Fornitore: Thermo Fisher Scientific
Attenzione

Synonyms: 2-Hydroxymethyl-3,4-dihydro-2H-pyran, DHP Linker

H56124.03 H56124.06
H56124.03EA 38.5 EUR
H56124.03 H56124.06
3,4-Dihydro-2H-pyran-2-methanol 96%
3,4-Dihydro-2H-pyran-2-methanol

Purity: 96% CAS No.: 3749-36-8 Molecular Formula: C6H10O2 Formula Weight: 114.14 Storage Temperature: 2°C to 8°C Flash Point: 89°(192°F) Refractive Index: 1.4780 Boiling Point: 92-93°/25mm MDL No.: MFCD00051507

Formula: C₆H₁₀O₂
Punto di ebollizione: 92…93 °C (25 mmHg)
Densità: 1,101
Flash Pt: 89 °C (192 °F)
MDL Number: MFCD00051507
CAS Number: 3749-36-8
EINECS: 223-150-0

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR