Stampa…

3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate ≥98.0% (by HPLC)

Fornitore: Thermo Fisher Scientific
Pericolo

Synonyms: 2-Hydrazono-3-methyl-2,3-dihydrobenzo[d]thiazole hydrochloride hydrate, 2-Hydrazono-3-methylbenzothiazoline hydrochloride

H56061.06 H56061.18
H56061.06EA 61.4 EUR
H56061.06 H56061.18
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate ≥98.0% (by HPLC)
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate

It is used as a reagent for the spectrophotometric determination of aliphatic aldehydes and for the determination of hexosamines in glycosaminoglycans. It is Incorporated into a new peroxidase color reaction. It is also a reagent for the spectrophotometric determination of traces of selenium (IV) in environmental samples.


Attenzione: Incompatible with oxidizing agents. Store at room temperature.

Formula: C₈H₉N₃S·H₂O·HCl
Peso molecolare: 233.714 g/mol
Punto di fusione: 270...274 °C
MDL Number: MFCD00149370
CAS Number: 38894-11-0
EINECS: 238-428-7
UN: 2811
ADR: 6.1 III

Visualizza i codici

Risultati del test di specificazione

Assay (HPLC) 98.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR