Stampa…

1-Ethynylpyrene ≥98%

Fornitore: Thermo Fisher Scientific

H37327.MDEA 29.6 EUR
H37327.MD
1-Ethynylpyrene ≥98%
1-Ethynylpyrene

Purity: 98+% CAS No.: 34993-56-1 Molecular Formula: C18H10 Physical Form: Powder Appearance: Yellow Melting Point: 113-116° Beilstein Reference: 2365008 MDL No.: MFCD02093933

Formula: C₁₈H₁₀
Punto di fusione: 113…116 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD02093933
CAS Number: 34993-56-1

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR