Stampa…

cis-4-Aminocyclohexanecarboxylic acid 97%

Fornitore: Thermo Fisher Scientific
Attenzione

H27567.03 H27567.06
H27567.03EA 54.8 EUR
H27567.03 H27567.06
cis-4-Aminocyclohexanecarboxylic acid 97%
cis-4-Aminocyclohexanecarboxylic acid

Purity: 97% CAS No.: 3685-23-2 Molecular Formula: C7H13NO2 Formula Weight: 143.19 Melting Point: 299-301° MDL No.: MFCD00191730

Formula: C₇H₁₃NO₂
Punto di fusione: 299…301 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00191730
CAS Number: 3685-23-2

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR