Stampa…
Jerrican, quadrato e rettangolare, HDPE, Corning® Gosselin™
Fornitore: Corning
JCP10-02
GOSSJCP10-02EA 122 EUR
GOSSJCP10-02
Jerrican, quadrato e rettangolare, HDPE, Corning® Gosselin™
Bottiglie Taniche
HDPE, translucent.
  • Good chemical resistance
  • Temperature resistant from −40 to +110 °C
Universal jerricans for industry, trade and laboratories. Ideal for packaging, storage and transportation.