Stampa…
Test tubes with conical winged base, Corning® Gosselin™
Fornitore: Corning
CC21-01
GOSSCC21-01EA 227 EUR
GOSSCC21-01
Test tubes with conical winged base, Corning® Gosselin™
Tubi Provette
Polystyrene, natural, without cap, aseptic.
  • Ideal for centrifugation of samples
  • Tubes with caps are leakproof
  • Can withstand centrifugation to 3000 g