Stampa…

Ematossilina Gill No. 1, Sigma-Aldrich®

Fornitore: SIGMA ALDRICH MICROSCOPY
Attenzione

GHS116-500ML
GHS116-500MLEA 64.4 EUR
GHS116-500ML
Ematossilina Gill No. 1, Sigma-Aldrich®
Ematossilina (soluzione colorante)

Gill No. 1 formulation is used as a progressive cytology stain. Used with hematoxylin and eosin staining.

Formula: C₁₆H₁₄O₆
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00078111
CAS Number: 517-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea092c5c7a1197808ab2551b9dd69b8488526407&vid=1cd118d1d860bc563bb6193c7444d67c5e272273&prid=web&clid=SAPDAM IVDLogo

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR